siRNA Details
Gene Symbol: HMGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145205.5 | NP_660206.2 | 2 | 2200 | ||
NR_033264.1 | 2 | 485 | ||||
GenBank mRNA | AK097707.1 | - | 2 | 2201 | ||
AK130859.1 | - | 1 | 619 | |||
AK310465.1 | - | 2 | 485 | |||
AK311985.1 | - | 1 | 586 | |||
BC021180.2 | - | 1 | 570 | 5266418 | ||
BC070148.1 | - | 2 | 485 | 6617009 | ||
BI827031.1 | - | 1 | 572 | |||
DQ207367.1 | - | 1 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
