siRNA Details
Gene Symbol: CLYBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206808.3 | NP_996531.1 | 3 | 330 | ||
NR_104592.1 | 3 | 330 | ||||
XM_005254030.2 | XP_005254087.1 | 3 | 309 | |||
XM_005254031.2 | XP_005254088.1 | 3 | 309 | |||
XM_006719915.3 | XP_006719978.1 | 3 | 309 | |||
XM_011521051.2 | XP_011519353.1 | 3 | 309 | |||
XM_011521052.2 | XP_011519354.1 | 3 | 309 | |||
XM_017020402.1 | XP_016875891.1 | 3 | 309 | |||
XM_017020403.1 | XP_016875892.1 | 3 | 1130 | |||
XM_017020404.1 | XP_016875893.1 | 3 | 1279 | |||
GenBank mRNA | AF428253.1 | - | 3 | 298 | ||
AK095506.1 | - | 3 | 307 | |||
AK300510.1 | - | 3 | 325 | |||
BC034360.1 | - | 3 | 312 | 4750050 | ||
DA621336.1 | - | 3 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
