siRNA Details
Gene Symbol: NLRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277126.1 | NP_001264055.1 | 5 | 2558 | ||
NM_001277129.1 | NP_001264058.1 | 5 | 2555 | |||
NM_144687.3 | NP_653288.1 | 5 | 2555 | |||
XM_011527479.1 | XP_011525781.1 | 5 | 2558 | |||
XM_011527480.1 | XP_011525782.1 | 5 | 2558 | |||
XM_011527482.1 | XP_011525784.1 | 5 | 2558 | |||
XM_017027460.1 | XP_016882949.1 | 5 | 2558 | |||
XM_017027461.1 | XP_016882950.1 | 5 | 2558 | |||
XM_017027462.1 | XP_016882951.1 | 4 | 2384 | |||
XM_017027463.1 | XP_016882952.1 | 6 | 2331 | |||
XM_017027464.1 | XP_016882953.1 | 5 | 2129 | |||
XM_017027465.1 | XP_016882954.1 | 6 | 2339 | |||
XM_017027466.1 | XP_016882955.1 | 6 | 2336 | |||
XM_017027467.1 | XP_016882956.1 | 5 | 2131 | |||
GenBank mRNA | AF231021.1 | - | 3 | 1483 | ||
AK095460.1 | - | 5 | 2470 | |||
AK290772.1 | - | 5 | 2337 | |||
AY095146.1 | - | 5 | 2502 | |||
AY116204.1 | - | 5 | 2549 | |||
AY116205.1 | - | 5 | 2549 | |||
AY116206.1 | - | 5 | 2549 | |||
AY116207.1 | - | 5 | 2549 | |||
AY154467.1 | - | 5 | 2502 | |||
BC028069.1 | - | 5 | 2445 | 5212737 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
