siRNA Details
Gene Symbol: FANCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018113.2 | NP_001018123.1 | 3 | 651 | ||
NM_001324162.1 | NP_001311091.1 | 3 | 630 | |||
NM_152633.3 | NP_689846.1 | 2 | 530 | |||
XM_011545470.2 | XP_011543772.1 | 3 | 837 | |||
XM_017029355.1 | XP_016884844.1 | 3 | 729 | |||
XM_017029356.1 | XP_016884845.1 | 3 | 837 | |||
GenBank mRNA | AK091383.1 | - | 2 | 530 | ||
BC043596.1 | - | 3 | 634 | |||
BC136558.1 | - | 3 | 633 | 9020548 | ||
BC136560.1 | - | 2 | 512 | 9020550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
