siRNA Details
Gene Symbol: GTPBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128855.2 | NP_001122327.1 | 9 | 2417 | ||
NM_001195422.1 | NP_001182351.1 | 9 | 2473 | |||
NM_032620.3 | NP_116009.2 | 9 | 2480 | |||
NM_133644.3 | NP_598399.2 | 8 | 2576 | |||
GenBank mRNA | AK027606.1 | - | 9 | 2433 | ||
AK091035.1 | - | 9 | 2798 | |||
AY078988.1 | - | 9 | 2433 | |||
BC008924.2 | - | 7 | 1963 | |||
BC019261.1 | - | 9 | 2428 | 3501739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
