siRNA Details
Gene Symbol: GTPBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128855.2 | NP_001122327.1 | 9 | 1608 | ||
NM_001195422.1 | NP_001182351.1 | 9 | 1664 | |||
NM_032620.3 | NP_116009.2 | 9 | 1671 | |||
NM_133644.3 | NP_598399.2 | 8 | 1767 | |||
GenBank mRNA | AF360742.1 | - | 9 | 1633 | ||
AF360743.1 | - | 9 | 1570 | |||
AF360744.1 | - | 9 | 1550 | |||
AK027606.1 | - | 9 | 1618 | |||
AK291929.1 | - | 9 | 1615 | |||
AK297953.1 | - | 9 | 1646 | |||
AY078987.1 | - | 8 | 1729 | |||
AY078988.1 | - | 9 | 1633 | |||
BC008924.2 | - | 7 | 1148 | |||
BC017207.2 | - | 9 | 1603 | 3634066 | ||
BC019261.1 | - | 9 | 1612 | 3501739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
