siRNA Details
Gene Symbol: GTF2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163636.2 | NP_001157108.1 | 34 | 3859 | ||
NM_001518.4 | NP_001509.3 | 33 | 3802 | |||
NM_032999.3 | NP_127492.1 | 35 | 3925 | |||
NM_033000.3 | NP_127493.1 | 34 | 3865 | |||
NM_033001.3 | NP_127494.1 | 34 | 3862 | |||
GenBank mRNA | AF015553.1 | - | 33 | 3766 | ||
AF035737.1 | - | 35 | 3832 | |||
AF038967.1 | - | 34 | 3772 | |||
AF038968.1 | - | 34 | 3769 | |||
AF038969.1 | - | 33 | 3709 | |||
AK093663.1 | - | 4 | 2349 | |||
AK096095.1 | - | 1 | 1198 | |||
BC070484.1 | - | 33 | 3745 | 5815493 | ||
BC099907.1 | - | 34 | 3791 | 5263270 |
Gene Symbol: GTF2IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002206.3 | 24 | 2984 | |||
GenBank mRNA | DQ786291.1 | - | 4 | 711 |
Gene Symbol: GTF2IP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003580.2 | 24 | 2993 | |||
GenBank mRNA | AK057750.1 | - | 1 | 958 | ||
AK098799.1 | - | 6 | 1660 | |||
CN368586.1 | - | 1 | 224 | |||
CX762709.1 | - | 2 | 589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
