siRNA Details
Gene Symbol: GTF2IP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003580.2 | 8, 9 | 1151 | |||
GenBank mRNA | CX873251.1 | - | 8, 9 | 761 | ||
DA067811.1 | - | 2, 3 | 133 | |||
DA510074.1 | - | 2, 3 | 99 |
Gene Symbol: GTF2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163636.2 | NP_001157108.1 | 18, 19 | 2017 | ||
NM_001518.4 | NP_001509.3 | 17, 18 | 1960 | |||
NM_032999.3 | NP_127492.1 | 19, 20 | 2083 | |||
NM_033000.3 | NP_127493.1 | 18, 19 | 2023 | |||
NM_033001.3 | NP_127494.1 | 18, 19 | 2020 | |||
GenBank mRNA | AF015553.1 | - | 17, 18 | 1921 | ||
AK292832.1 | - | 18, 19 | 2008 | |||
AK294935.1 | - | 20, 21 | 1917 | |||
BC070484.1 | - | 17, 18 | 1903 | 5815493 | ||
BC099907.1 | - | 18, 19 | 1949 | 5263270 | ||
KJ534859.1 | - | 16, 17 | 1551 | |||
KJ534860.1 | - | 18, 19 | 1674 | |||
KJ534861.1 | - | 17, 18 | 1614 | |||
KJ534862.1 | - | 18, 19 | 1674 | |||
KJ534863.1 | - | 17, 18 | 1611 | |||
U77948.1 | - | 16, 17 | 1557 | |||
Y14946.1 | - | 16, 17 | 1551 |
Gene Symbol: GTF2IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002206.3 | 8, 9 | 1144 | |||
GenBank mRNA | AK299175.1 | - | 8, 9 | 1128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
