siRNA Details
Gene Symbol: GTF2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163636.2 | NP_001157108.1 | 23 | 2465 | ||
NM_001518.4 | NP_001509.3 | 22 | 2408 | |||
NM_032999.3 | NP_127492.1 | 24 | 2531 | |||
NM_033000.3 | NP_127493.1 | 23 | 2471 | |||
NM_033001.3 | NP_127494.1 | 23 | 2468 | |||
GenBank mRNA | AF015553.1 | - | 22 | 2369 | ||
AF035737.1 | - | 24 | 2438 | |||
AF038967.1 | - | 23 | 2378 | |||
AF038968.1 | - | 23 | 2375 | |||
AF038969.1 | - | 22 | 2315 | |||
AK292832.1 | - | 23 | 2456 | |||
AK294935.1 | - | 25 | 2365 | |||
BC070484.1 | - | 22 | 2351 | 5815493 | ||
BC099907.1 | - | 23 | 2397 | 5263270 | ||
KJ534859.1 | - | 21 | 1999 | |||
KJ534860.1 | - | 23 | 2122 | |||
KJ534861.1 | - | 22 | 2062 | |||
KJ534862.1 | - | 23 | 2122 | |||
KJ534863.1 | - | 22 | 2059 | |||
U77948.1 | - | 21 | 2005 | |||
Y14946.1 | - | 21 | 1999 |
Gene Symbol: GTF2IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002206.3 | 13 | 1592 | |||
GenBank mRNA | AK299175.1 | - | 13 | 1576 |
Gene Symbol: GTF2IP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003580.2 | 13 | 1599 | |||
GenBank mRNA | BM558161.1 | - | 2 | 123 | ||
DA740051.1 | - | 2 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
