siRNA Details
Gene Symbol: GRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198182.2 | NP_937825.2 | 5, 6 | 928 | ||
XM_005246159.3 | XP_005246216.1 | 4, 5 | 814 | |||
XM_006711882.2 | XP_006711945.1 | 5, 6 | 928 | |||
XM_006711884.3 | XP_006711947.1 | 4, 5 | 807 | |||
XM_011510343.2 | XP_011508645.1 | 5, 6 | 899 | |||
XM_017003900.1 | XP_016859389.1 | 5, 6 | 1001 | |||
XM_017003901.1 | XP_016859390.1 | 5, 6 | 1001 | |||
GenBank mRNA | AF198489.1 | - | 5, 6 | 866 | ||
AF411210.1 | - | 1, 2 | 283 | |||
AK293855.1 | - | 5, 6 | 975 | |||
AK296982.1 | - | 5, 6 | 1006 | |||
AK312950.1 | - | 5, 6 | 898 | |||
BC067519.1 | - | 5, 6 | 809 | 6971723 | ||
BC067520.1 | - | 5, 6 | 809 | 6971724 | ||
BC067521.1 | - | 5, 6 | 809 | 6971725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
