siRNA Details
Gene Symbol: GRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198182.2 | NP_937825.2 | 4 | 740 | ||
XM_005246159.3 | XP_005246216.1 | 4 | 703 | |||
XM_006711882.2 | XP_006711945.1 | 4 | 740 | |||
XM_006711884.3 | XP_006711947.1 | 4 | 696 | |||
XM_011510343.2 | XP_011508645.1 | 4 | 711 | |||
XM_017003900.1 | XP_016859389.1 | 4 | 813 | |||
XM_017003901.1 | XP_016859390.1 | 4 | 813 | |||
GenBank mRNA | AF198489.1 | - | 4 | 678 | ||
AF411210.1 | - | 1 | 172 | |||
AK293855.1 | - | 5 | 864 | |||
AK296982.1 | - | 4 | 818 | |||
AK312950.1 | - | 4 | 710 | |||
BC067519.1 | - | 4 | 621 | 6971723 | ||
CD559574.2 | - | 4 | 630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
