siRNA Details
Gene Symbol: TMEM64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008495.3 | NP_001008495.2 | 3 | 1853 | ||
NM_001146273.1 | NP_001139745.1 | 2 | 1697 | |||
GenBank mRNA | AK095472.1 | - | 1 | 64 | ||
AK123958.1 | - | 3 | 1547 | |||
AL834364.1 | - | 3 | 1558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
