siRNA Details
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004228.6 | NP_004219.3 | 12 | 1643 | ||
NM_017457.5 | NP_059431.1 | 13 | 1646 | |||
XM_006723472.2 | XP_006723535.1 | 12 | 1729 | |||
GenBank mRNA | AB209573.1 | - | 11 | 4594 | ||
BC004361.1 | - | 13 | 1461 | 3538580 | ||
BC038713.1 | - | 13 | 1503 | 5269719 | ||
U59752.1 | - | 9 | 944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
