siRNA Details
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004228.6 | NP_004219.3 | 10 | 1226 | ||
NM_017457.5 | NP_059431.1 | 11 | 1229 | |||
XM_006723472.2 | XP_006723535.1 | 10 | 1312 | |||
XM_006723473.2 | XP_006723536.1 | 11 | 1315 | |||
GenBank mRNA | AB209573.1 | - | 9 | 4177 | ||
AK292405.1 | - | 11 | 1086 | |||
BC004361.1 | - | 11 | 1044 | 3538580 | ||
BC038713.1 | - | 11 | 1086 | 5269719 | ||
U59752.1 | - | 7 | 527 | |||
U70728.1 | - | 11 | 1087 | |||
X99753.1 | - | 10 | 926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
