siRNA Details
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004228.6 | NP_004219.3 | 7 | 910 | ||
NM_017457.5 | NP_059431.1 | 7 | 910 | |||
XM_006723472.2 | XP_006723535.1 | 7 | 996 | |||
XM_006723473.2 | XP_006723536.1 | 7 | 996 | |||
GenBank mRNA | AB209573.1 | - | 7 | 742 | ||
AK091443.1 | - | 2 | 1360 | |||
AK292405.1 | - | 7 | 767 | |||
BC004361.1 | - | 7 | 725 | 3538580 | ||
BC038713.1 | - | 7 | 767 | 5269719 | ||
U59752.1 | - | 3 | 208 | |||
U70728.1 | - | 7 | 768 | |||
X99753.1 | - | 7 | 610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
