siRNA Details
Gene Symbol: KIAA0319L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024874.4 | NP_079150.3 | 3 | 886 | ||
XM_006710907.1 | XP_006710970.1 | 3 | 685 | |||
XM_006710909.1 | XP_006710972.1 | 3 | 777 | |||
XM_011542179.2 | XP_011540481.1 | 3 | 1017 | |||
XM_011542180.1 | XP_011540482.1 | 3 | 855 | |||
XM_017002366.1 | XP_016857855.1 | 3 | 855 | |||
XM_017002367.1 | XP_016857856.1 | 3 | 1017 | |||
XM_017002368.1 | XP_016857857.1 | 3 | 685 | |||
XM_017002369.1 | XP_016857858.1 | 3 | 777 | |||
XM_017002370.1 | XP_016857859.1 | 3 | 855 | |||
XM_017002371.1 | XP_016857860.1 | 3 | 855 | |||
XM_017002372.1 | XP_016857861.1 | 3 | 777 | |||
XM_017002373.1 | XP_016857862.1 | 3 | 855 | |||
XM_017002375.1 | XP_016857864.1 | 1 | 218 | |||
XM_017002376.1 | XP_016857865.1 | 1 | 218 | |||
GenBank mRNA | AF275679.2 | - | 3 | 835 | ||
AF289597.1 | - | 1 | 451 | |||
AK302428.1 | - | 3 | 896 | |||
AK314605.1 | - | 3 | 834 | |||
AK316252.1 | - | 3 | 1014 | |||
AY163234.1 | - | 3 | 835 | |||
BC014530.1 | - | 3 | 818 | 3843301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
