siRNA Details
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194998.1 | NP_001181927.1 | 20 | 3417 | ||
NM_014985.3 | NP_055800.2 | 20 | 3417 | |||
XM_006720437.3 | XP_006720500.1 | 20 | 3602 | |||
XM_011521373.2 | XP_011519675.1 | 20 | 4170 | |||
XM_011521374.2 | XP_011519676.1 | 20 | 4169 | |||
XM_011521375.2 | XP_011519677.1 | 20 | 4169 | |||
XM_011521378.2 | XP_011519680.1 | 20 | 4169 | |||
XM_011521379.2 | XP_011519681.1 | 20 | 4168 | |||
XM_011521381.2 | XP_011519683.1 | 8 | 2330 | |||
XM_017022014.1 | XP_016877503.1 | 13 | 2363 | |||
XM_017022015.1 | XP_016877504.1 | 8 | 1519 | |||
GenBank mRNA | AB020719.1 | - | 19 | 3022 | ||
BC117182.1 | - | 20 | 3258 | 40125733 | ||
BX648822.1 | - | 20 | 3344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
