siRNA Details
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194998.1 | NP_001181927.1 | 11 | 1581 | ||
NM_014985.3 | NP_055800.2 | 11 | 1581 | |||
XM_006720437.3 | XP_006720500.1 | 11 | 1766 | |||
XM_011521373.2 | XP_011519675.1 | 11 | 2334 | |||
XM_011521374.2 | XP_011519676.1 | 11 | 2333 | |||
XM_011521375.2 | XP_011519677.1 | 11 | 2333 | |||
XM_011521378.2 | XP_011519680.1 | 11 | 2333 | |||
XM_011521379.2 | XP_011519681.1 | 11 | 2332 | |||
XM_017022014.1 | XP_016877503.1 | 4 | 527 | |||
XM_017022016.1 | XP_016877505.1 | 11 | 2336 | |||
GenBank mRNA | AB020719.1 | - | 10 | 1186 | ||
AK303805.1 | - | 11 | 1504 | |||
BC069186.1 | - | 11 | 1477 | |||
BC107721.1 | - | 11 | 1585 | |||
BC117182.1 | - | 11 | 1422 | 40125733 | ||
BX648822.1 | - | 11 | 1508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
