siRNA Details
Gene Symbol: FAXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032385.4 | NP_115761.2 | 8 | 1033 | ||
XM_005268358.2 | XP_005268415.1 | 5 | 678 | |||
XM_006714753.2 | XP_006714816.1 | 9 | 1408 | |||
XM_017008965.1 | XP_016864454.1 | 9 | 1433 | |||
XM_017008966.1 | XP_016864455.1 | 6 | 708 | |||
GenBank mRNA | AF159165.1 | - | 1 | 252 | ||
AK023820.1 | - | 8 | 1037 | |||
AK074245.1 | - | 2 | 178 | |||
AK127944.1 | - | 3 | 1580 | |||
AK295546.1 | - | 7 | 910 | |||
BC004506.2 | - | 6 | 730 | |||
BC007216.1 | - | 8 | 1001 | 2906244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
