siRNA Details
Gene Symbol: NFKBIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005474.2 | NP_001005474.1 | 9 | 1622 | ||
NM_031419.3 | NP_113607.1 | 8 | 1755 | |||
GenBank mRNA | AB037925.1 | - | 8 | 1688 | ||
AF548362.1 | - | 9 | 1619 | |||
AK054787.1 | - | 9 | 1491 | |||
AK091782.1 | - | 9 | 1389 | |||
BC014587.2 | - | 1401 | ||||
BC060800.1 | - | 8 | 1755 | 30344780 | ||
DQ224339.1 | - | 8 | 1544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
