siRNA Details
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 20 | 2447 | ||
NM_001278513.1 | NP_001265442.1 | 20 | 2591 | |||
NM_001278514.1 | NP_001265443.1 | 20 | 2259 | |||
NM_003589.2 | NP_003580.1 | 20 | 2273 | |||
XM_011537523.2 | XP_011535825.1 | 19 | 2807 | |||
GenBank mRNA | AF077188.1 | - | 20 | 2184 | ||
AK296700.1 | - | 12 | 1568 | |||
AL833355.1 | - | 19 | 2211 | |||
BC008308.1 | - | 20 | 2335 | 3537176 | ||
U58090.1 | - | 10 | 1311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
