siRNA Details
Gene Symbol: COX6B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144613.4 | NP_653214.2 | 5 | 691 | ||
GenBank mRNA | AK057427.1 | - | 8 | 1117 | ||
BC026123.2 | - | 5 | 692 | 4827471 | ||
BC064548.1 | - | 6 | 576 | |||
BC100899.2 | - | 4 | 528 | 40003591 | ||
BC100900.1 | - | 5 | 531 | 40003592 | ||
BC100901.1 | - | 4 | 528 | 40003593 | ||
BC100902.1 | - | 8 | 1136 | 40003594 | ||
BG717038.1 | - | 5 | 697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
