siRNA Details
Gene Symbol: MAGED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005332.1 | NP_001005332.1 | 3 | 606 | ||
NM_001005333.1 | NP_001005333.1 | 4 | 869 | |||
NM_006986.3 | NP_008917.3 | 3 | 701 | |||
XM_011530835.2 | XP_011529137.1 | 4 | 724 | |||
XM_017029978.1 | XP_016885467.1 | 3 | 616 | |||
XM_017029979.1 | XP_016885468.1 | 4 | 1302 | |||
XM_017029980.1 | XP_016885469.1 | 4 | 1120 | |||
GenBank mRNA | AF217963.1 | - | 3 | 651 | ||
AF258554.1 | - | 3 | 675 | |||
AF300328.1 | - | 3 | 652 | |||
AK074092.1 | - | 3 | 654 | |||
AK298574.1 | - | 4 | 457 | |||
AK314053.1 | - | 3 | 664 | |||
BC014070.2 | - | 4 | 832 | 4026470 | ||
BC032473.2 | - | 2 | 548 | 5216354 | ||
CD387872.1 | - | 3 | 690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
