siRNA Details
Gene Symbol: DKK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018057.1 | NP_001018067.1 | 4, 5 | 587 | ||
NM_013253.4 | NP_037385.2 | 5, 6 | 755 | |||
NM_015881.5 | NP_056965.3 | 5, 6 | 769 | |||
XM_006718178.2 | XP_006718241.1 | 5, 6 | 582 | |||
XM_017017553.1 | XP_016873042.1 | 5, 6 | 716 | |||
XM_017017554.1 | XP_016873043.1 | 5, 6 | 734 | |||
XM_017017555.1 | XP_016873044.1 | 5, 6 | 582 | |||
GenBank mRNA | AB033421.1 | - | 4, 5 | 548 | ||
AB034203.1 | - | 5, 6 | 727 | |||
AB057591.1 | - | 5, 6 | 755 | |||
AF177396.1 | - | 4, 5 | 567 | |||
AK092979.1 | - | 5, 6 | 546 | |||
AK225156.1 | - | 4, 5 | 539 | |||
AK289897.1 | - | 5, 6 | 596 | |||
AK295441.1 | - | 3, 4 | 447 | |||
AK295539.1 | - | 5, 6 | 690 | |||
AL535720.3 | - | 4, 5 | 586 | |||
AY358378.1 | - | 5, 6 | 682 | |||
AY587550.1 | - | 4, 5 | 530 | |||
BC007660.2 | - | 5, 6 | 768 | 3347453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
