siRNA Details
Gene Symbol: LOC643549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097925.1 | - | 1 | 400 | ||
BC133006.1 | - | 1 | 337 | |||
BC133008.1 | - | 1 | 337 |
Gene Symbol: LOC107985388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028186.1 | XP_016883675.1 | 2 | 759 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
