siRNA Details
Gene Symbol: CLUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289036.1 | NP_001275965.1 | 4 | 472 | ||
NM_001318522.1 | NP_001305451.1 | 2 | 188 | |||
NM_014410.4 | NP_055225.1 | 2 | 227 | |||
NM_199167.1 | NP_954636.1 | 2 | 153 | |||
XM_005258103.3 | XP_005258160.1 | 4 | 377 | |||
XM_006722322.2 | XP_006722385.1 | 3 | 345 | |||
XM_011525648.2 | XP_011523950.1 | 4 | 420 | |||
XM_011525649.1 | XP_011523951.1 | 4 | 420 | |||
XM_011525651.2 | XP_011523953.1 | 3 | 1142 | |||
XM_011525652.1 | XP_011523954.1 | 4 | 496 | |||
XM_011525653.2 | XP_011523955.1 | 2 | 117 | |||
XM_017025707.1 | XP_016881196.1 | 5 | 1882 | |||
GenBank mRNA | AF395889.1 | - | 2 | 142 | ||
AK299319.1 | - | 4 | 447 | |||
AL712819.1 | - | 2 | 153 | |||
AL713125.1 | - | 3 | 360 | |||
BC025381.2 | - | 2 | 227 | 4821299 | ||
D63813.1 | - | 2 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
