siRNA Details
Gene Symbol: CLUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289036.1 | NP_001275965.1 | 11 | 2107 | ||
NM_001318522.1 | NP_001305451.1 | 9 | 1823 | |||
NM_014410.4 | NP_055225.1 | 9 | 1862 | |||
NM_199167.1 | NP_954636.1 | 9 | 1788 | |||
XM_005258103.3 | XP_005258160.1 | 11 | 2012 | |||
XM_006722322.2 | XP_006722385.1 | 10 | 1980 | |||
XM_011525649.1 | XP_011523951.1 | 11 | 2055 | |||
XM_011525651.2 | XP_011523953.1 | 10 | 2777 | |||
XM_011525652.1 | XP_011523954.1 | 11 | 2131 | |||
XM_017025707.1 | XP_016881196.1 | 12 | 3517 | |||
GenBank mRNA | AK299319.1 | - | 11 | 2082 | ||
BC025381.2 | - | 9 | 1862 | 4821299 | ||
BX108184.1 | - | 2 | 526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
