siRNA Details
Gene Symbol: PPP6R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014931.3 | NP_055746.3 | 11, 12 | 1597 | ||
GenBank mRNA | AB029038.2 | - | 11, 12 | 1922 | ||
BC002799.2 | - | 10, 11 | 1199 | |||
BC068014.1 | - | 10, 11 | 1261 | |||
BC094753.1 | - | 11, 12 | 1459 | 4838365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
