siRNA Details
Gene Symbol: MAML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018717.4 | NP_061187.2 | 5 | 4025 | ||
GenBank mRNA | AK123604.1 | - | 5 | 3343 | ||
AK296891.1 | - | 4 | 1317 | |||
AL359614.1 | - | 1 | 389 | |||
BC137130.1 | - | 5 | 3338 | 9021127 | ||
BC137131.1 | - | 5 | 3338 | 9021128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
