siRNA Details
Gene Symbol: MAML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018717.4 | NP_061187.2 | 5 | 3356 | ||
GenBank mRNA | AK123604.1 | - | 5 | 2674 | ||
AK296891.1 | - | 4 | 648 | |||
BC137130.1 | - | 5 | 2669 | 9021127 | ||
BC137131.1 | - | 5 | 2669 | 9021128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
