siRNA Details
Gene Symbol: FAM110C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077710.2 | NP_001071178.2 | 1 | 859 | ||
XM_011510372.2 | XP_011508674.1 | 1 | 861 | |||
XM_017004689.1 | XP_016860178.1 | 2 | 1053 | |||
XM_017004690.1 | XP_016860179.1 | 2 | 801 | |||
XM_017004691.1 | XP_016860180.1 | 1 | 861 | |||
XM_017004692.1 | XP_016860181.1 | 1 | 861 | |||
GenBank mRNA | AK302544.1 | - | 1 | 800 | ||
DQ431183.1 | - | 1 | 656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
