siRNA Details
Gene Symbol: USP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080481.1 | NP_001073950.1 | 7 | 1218 | ||
XM_005267169.3 | XP_005267226.1 | 7 | 821 | |||
XM_005267170.4 | XP_005267227.1 | 7 | 790 | |||
XM_005267172.3 | XP_005267229.1 | 7 | 790 | |||
XM_005267175.3 | XP_005267232.1 | 7 | 789 | |||
XM_011536200.2 | XP_011534502.1 | 7 | 2048 | |||
XM_017011382.1 | XP_016866871.1 | 7 | 1215 | |||
XM_017011383.1 | XP_016866872.1 | 7 | 741 | |||
XM_017011384.1 | XP_016866873.1 | 7 | 718 | |||
XM_017011385.1 | XP_016866874.1 | 7 | 789 | |||
XM_017011386.1 | XP_016866875.1 | 7 | 789 | |||
GenBank mRNA | AJ583819.2 | - | 6 | 685 | ||
AK310643.1 | - | 7 | 722 | |||
BC005991.1 | - | 7 | 736 | |||
BC150648.2 | - | 7 | 785 | 9057019 | ||
BC157838.1 | - | 7 | 785 | 9057057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
