siRNA Details
Gene Symbol: PHF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278221.2 | NP_001265150.1 | 7 | 1312 | ||
NM_001321126.1 | NP_001308055.1 | 7 | 1271 | |||
NM_001321127.1 | NP_001308056.1 | 7 | 1404 | |||
NM_016483.6 | NP_057567.3 | 7 | 1312 | |||
XM_011533826.2 | XP_011532128.1 | 7 | 1262 | |||
GenBank mRNA | AF151060.2 | - | 6 | 479 | ||
AK223531.1 | - | 6 | 529 | |||
AK292221.1 | - | 7 | 1118 | |||
AL137671.1 | - | 2 | 230 | |||
AY014283.2 | - | 7 | 886 | |||
BC022002.1 | - | 7 | 1171 | 4822315 | ||
BU853281.1 | - | 2 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
