siRNA Details
Gene Symbol: PHF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278221.2 | NP_001265150.1 | 10 | 1961 | ||
NM_001321126.1 | NP_001308055.1 | 11 | 2037 | |||
NM_001321127.1 | NP_001308056.1 | 11 | 2170 | |||
NM_016483.6 | NP_057567.3 | 11 | 2078 | |||
GenBank mRNA | AF151060.2 | - | 9 | 1126 | ||
AK223531.1 | - | 10 | 1295 | |||
AK292221.1 | - | 11 | 1884 | |||
AY014283.2 | - | 11 | 1652 | |||
BC022002.1 | - | 11 | 1937 | 4822315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
