siRNA Details
Gene Symbol: GRID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017551.2 | NP_060021.1 | 10 | 1498 | ||
XM_011539720.2 | XP_011538022.1 | 10 | 1495 | |||
GenBank mRNA | AB033046.1 | - | 7 | 745 | ||
AK127168.1 | - | 10 | 1483 | |||
AK293104.1 | - | 10 | 1456 | |||
AK302192.1 | - | 3 | 270 | |||
BC039263.1 | - | 10 | 1498 | 5273869 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
