siRNA Details
Gene Symbol: RAET1E-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045126.1 | 2 | 1163 | |||
NR_045127.1 | 3 | 447 | ||||
GenBank mRNA | BC016788.1 | - | 2 | 1155 | ||
DB521065.1 | - | 3 | 392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
