siRNA Details
Gene Symbol: NLRP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127255.1 | NP_001120727.1 | 11 | 3297 | ||
NM_139176.3 | NP_631915.2 | 11 | 3213 | |||
NM_206828.3 | NP_996611.2 | 10 | 3126 | |||
XM_006723075.3 | XP_006723138.1 | 12 | 3532 | |||
XM_006723076.3 | XP_006723139.1 | 11 | 3495 | |||
XM_011526596.2 | XP_011524898.1 | 13 | 3739 | |||
XM_011526599.2 | XP_011524901.1 | 13 | 3750 | |||
XM_011526601.2 | XP_011524903.1 | 12 | 3568 | |||
GenBank mRNA | AF464765.1 | - | 9 | 3096 | ||
AK298630.1 | - | 9 | 2367 | |||
AK302655.1 | - | 13 | 3708 | |||
AY154462.1 | - | 10 | 3120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
