siRNA Details
Gene Symbol: KRT36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003771.4 | NP_003762.1 | 7 | 1568 | ||
GenBank mRNA | BC043581.1 | - | 8 | 1631 | 5227645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
