siRNA Details
Gene Symbol: SCFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283031.1 | NP_001269960.1 | 2 | 208 | ||
NM_001283032.1 | NP_001269961.1 | 2 | 208 | |||
NM_001283033.1 | NP_001269962.1 | 2 | 208 | |||
NM_016106.3 | NP_057190.2 | 2 | 208 | |||
XM_005267469.2 | XP_005267526.1 | 2 | 206 | |||
XM_011536586.2 | XP_011534888.1 | 2 | 205 | |||
XM_011536587.2 | XP_011534889.1 | 2 | 205 | |||
XM_011536588.2 | XP_011534890.1 | 2 | 205 | |||
XM_011536589.1 | XP_011534891.1 | 2 | 205 | |||
XM_017021113.1 | XP_016876602.1 | 2 | 205 | |||
GenBank mRNA | AB020724.2 | - | 2 | 143 | ||
AF086916.1 | - | 2 | 131 | |||
AF092139.1 | - | 2 | 124 | |||
AF110646.1 | - | 2 | 119 | |||
AK222820.1 | - | 2 | 127 | |||
AK290410.1 | - | 2 | 143 | |||
AK299222.1 | - | 2 | 135 | |||
AK301406.1 | - | 2 | 143 | |||
AK307960.1 | - | 2 | 143 | |||
AK308587.1 | - | 2 | 151 | |||
BC017734.1 | - | 2 | 123 | 4420306 | ||
BU198450.1 | - | 2 | 149 | |||
CA454055.1 | - | 2 | 134 | |||
CD697934.1 | - | 2 | 172 | |||
DA151789.1 | - | 2 | 206 | |||
DN993082.1 | - | 2 | 130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
