siRNA Details
Gene Symbol: MRGPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039165.2 | NP_001034254.2 | 2 | 494 | ||
GenBank mRNA | AK293940.1 | - | 2 | 494 | ||
AY255572.1 | - | 1 | 16 | |||
BC104889.1 | - | 1 | 218 | 8143892 | ||
BC112203.1 | - | 1 | 230 | 8327671 | ||
DC309261.1 | - | 2 | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
