siRNA Details
Gene Symbol: APOBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018690.3 | NP_061160.3 | 2 | 2855 | ||
GenBank mRNA | AF141332.1 | - | 3 | 2805 | ||
AF141334.1 | - | 3 | 2805 | |||
AK025123.1 | - | 1 | 707 | |||
AK075085.1 | - | 1 | 530 | |||
BC119786.1 | - | 3 | 2805 | 40008686 | ||
BC119788.1 | - | 3 | 2600 | 40008685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
