siRNA Details
Gene Symbol: PLCXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005473.2 | NP_001005473.1 | 3 | 7021 | ||
GenBank mRNA | AK124638.1 | - | 2 | 3157 | ||
BX648329.1 | - | 4 | 6832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
