siRNA Details
Gene Symbol: ZNF623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082480.2 | NP_001075949.1 | 2 | 1143 | ||
NM_001261843.1 | NP_001248772.1 | 2 | 1022 | |||
NM_014789.3 | NP_055604.3 | 1 | 1003 | |||
XM_006716708.3 | XP_006716771.1 | 2 | 1075 | |||
GenBank mRNA | AB014528.1 | - | 1 | 1127 | ||
AK294695.1 | - | 2 | 1015 | |||
BC067516.1 | - | 1 | 1003 | 7002215 | ||
BC067517.1 | - | 1 | 1003 | 7002216 | ||
BC067518.1 | - | 1 | 1003 | 7002218 | ||
BC075056.2 | - | 1 | 952 | 30915366 | ||
BC075057.2 | - | 1 | 950 | 30915582 | ||
BC112899.1 | - | 1 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
