siRNA Details
Gene Symbol: HIST1H3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003533.2 | NP_003524.1 | 1 | 231 | ||
GenBank mRNA | BC066884.1 | - | 1 | 231 | 7002077 | |
BC067493.1 | - | 1 | 231 | 7002079 | ||
BC069305.1 | - | 1 | 231 | 7262745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
