siRNA Details
Gene Symbol: OR2T29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004694.2 | NP_001004694.2 | 1 | 925 |
Gene Symbol: OR2T5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004697.1 | NP_001004697.1 | 1 | 925 | ||
GenBank mRNA | BC140747.1 | - | 1 | 930 | 9021615 | |
BC140915.1 | - | 1 | 950 | 9056727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
