siRNA Details
Gene Symbol: SYNM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015286.5 | NP_056101.5 | 4 | 2043 | ||
NM_145728.2 | NP_663780.2 | 4 | 2043 | |||
XM_017022035.1 | XP_016877524.1 | 4 | 2054 | |||
GenBank mRNA | AB002351.2 | - | 4 | 2081 | ||
AF359284.1 | - | 4 | 2034 | |||
AJ310521.1 | - | 4 | 2037 | |||
AJ310522.1 | - | 4 | 2037 | |||
AL833262.1 | - | 4 | 1534 | |||
BC151243.1 | - | 4 | 2088 | 8860516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
