siRNA Details
Gene Symbol: TCEAL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006612.1 | NP_001006613.1 | 3 | 838 | ||
NM_001006613.1 | NP_001006614.1 | 2 | 762 | |||
NM_001006614.1 | NP_001006615.1 | 2 | 759 | |||
NM_016303.2 | NP_057387.1 | 3 | 841 | |||
GenBank mRNA | AF125535.1 | - | 3 | 799 | ||
AL833152.1 | - | 1 | 1705 | |||
BC023544.2 | - | 3 | 792 | 4122244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
