siRNA Details
Gene Symbol: LOC729652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135198.1 | 1 | 684 | |||
GenBank mRNA | AL834536.1 | - | 1 | 684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
