siRNA Details
Gene Symbol: COA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008215.2 | NP_001008216.1 | 2 | 273 | ||
GenBank mRNA | AK131034.1 | - | 2 | 270 | ||
BC047722.1 | - | 2 | 247 | 5748383 | ||
DA214157.1 | - | 2 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
