siRNA Details
Gene Symbol: ZNF92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287532.1 | NP_001274461.1 | 3 | 2796 | ||
NM_001287533.1 | NP_001274462.1 | 5 | 3014 | |||
NM_001287534.1 | NP_001274463.1 | 2 | 2669 | |||
NM_007139.3 | NP_009070.2 | 3 | 2765 | |||
NM_152626.3 | NP_689839.1 | 4 | 2892 | |||
GenBank mRNA | AK091618.1 | - | 4 | 2810 | ||
AK093566.1 | - | 1 | 1668 | |||
BC036439.1 | - | 3 | 2688 | 4817733 | ||
BC040594.1 | - | 4 | 2834 | 5269950 | ||
BX647875.1 | - | 2 | 2613 | |||
EU668348.1 | - | 4 | 2836 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
